Polyethylene glycol 8,000 Biotechnology
Leverantör: VWR Chemicals
Varning
Synonyms: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
0159-1KGEA 3075 SEK
0159-1KG 0159-2.5KG 0159-500G
Polyethylene glycol 8,000 Biotechnology
Polyethylene glycol 8,000
Polyethylene glycol is used for the isolation of plasmid DNA and the precipitation of phage.
Formel: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Kokpunkt: >250 °C
Densitet: 1.21 g/cm³ (20 °C)
Flampunkt: >250 °C
Storage Temperature: Omgivande
MDL Number: MFCD01779615
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e4f2ac7d7b08ad5e6814488bdedf96bd458a77&vid=93a10b0bbb73df577d5985efd8329219e5fe1647&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
DNase None Detected
pH (5%, Water) @25 °C 4.5 – 7.5
RNase None Detected
Solidification Point 55 – 64 °C