Polyethylene glycol 2,000
Leverantör: Thermo Fisher Scientific
Synonyms: PEG 2000, Ethylene glycol polymer 2000, PEG 2,000, Ethylene glycol polymer average Mn 2000
B22181.0B B22181.30
B22181.0BEA 2175 SEK
B22181.0B B22181.30
Polyethylene glycol 2,000
Polyethylene glycol 2,000
CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Odour: Odourless pH: 5-7 Melting Point: 53-55° Merck Reference: 14,7568 MDL No.: MFCD01779611
Formel: H(OCH₂CH₂)nOH
MW: 2000 g/mol
Smältpunkt: 53…55 °C
Densitet: 1.21 g/cm³ (20 °C)
Flampunkt: 250
Storage Temperature: Omgivande
MDL Number: MFCD00081839
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568