Polyetylenglykol 1.500, skalor, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: PEG 1500, PEG 1,500
8.17005.5000 8.17005.9050
8.17005.5000EA 7000 SEK
8.17005.5000 8.17005.9050
Polyetylenglykol 1.500, skalor, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®
Polyetylenglykol 1.500
Formel: H(OCH₂CH₂)nOH
Kokpunkt: ∼250 °C (1013 hPa)
Smältpunkt: 44…48 °C
Densitet: 1,13 g/cm³ (20 °C)
Flampunkt: 260 °C
Storage Temperature: Omgivande
MDL Number: MFCD00081839
CAS nummer: 25322-68-3
EINECS: 500-038-2