Abies balsamea, Sigma-Aldrich®
Leverantör: SIGMA-ALDRICH MICROSCOPY
M-Clarity™ quality level = MQ100
C1795-100ML C1795-25ML
C1795-100MLEA 2910 SEK
C1795-100ML C1795-25ML
Abies balsamea, Sigma-Aldrich®
Abies balsamea
Densitet: 0,915 g/cm³ (20 °C)
Flampunkt: 62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00132800
CAS nummer: 8007-47-4
EINECS: 232-362-2
Merck Index: 13,00948