Abies balsamea för mikroskopi, Sigma-Aldrich®
Leverantör: Merck
Varning
M-Clarity™ quality level = MQ400
1.01691.0025 1.01691.0100
1.01691.0025EA 730 SEK
1.01691.0025 1.01691.0100
Abies balsamea för mikroskopi, Sigma-Aldrich®
Abies balsamea
Canada balsam is a commonly used mounting medium to prepare permanent slides for microscopy. It is produced from the resin of the balsam fir tree and its use can be combined with xylene-containing specimens. (Refractive index (20°C) 1.515 - 1.530) Canada balsam is an IVD product and CE registered.
Analysis Note
Appearance: nearly clear, yellow resinous
Refractive index (n 20/D): 1.515 - 1.530
Turbidity: ≤ 2 NTU
Acid value: 75 - 90
Loss on drying (120 °C): ≤ 35 %
Suitability for microscopy: passes test
Densitet: 0,915 g/cm³ (20 °C)
Flampunkt: 62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00132800
CAS nummer: 8007-47-4
EINECS: 232-362-2
UN: 1993
ADR: 3,III
Merck Index: 13,00948
IVDLogo