3-Mercapto-1,2-propanediol 90% 90%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol
125520250 125521000 125525000
ACRO125520250EA 770 SEK
ACRO125520250 ACRO125521000 ACRO125525000
3-Mercapto-1,2-propanediol 90% 90%
3-Mercapto-1,2-propanediol
Formel: C₃H₈O₂S
Kokpunkt: 118 °C (5 mmHg)
Densitet: 1,23 g/cm³
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004879
CAS nummer: 96-27-5
EINECS: 202-495-0
UN: 2810
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Viscous solution
Infrared spectrum Conforms
Titration Iodimetric ≥88 %
HPLC ≥96.0 %