L(+)-Lactic acid ∼90%, EMPROVE® EXPERT Ph. Eur., BP, E270 suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Fara
1.00366.2500 1.00366.9025
1.00366.2500EA 4600 SEK
1.00366.2500 1.00366.9025
L(+)-Lactic acid ∼90%, EMPROVE® EXPERT Ph. Eur., BP, E270 suitable for use as excipient, SAFC®
L(+)-Lactic acid
Formel: C₃H₆O₃
Densitet: 1,21 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00064266
CAS nummer: 79-33-4
EINECS: 201-196-2