L(+)-Lactic acid ∼85 - 90% aqueous solution
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
A17268.0E A17268.22 A17268.36
A17268.0EEA 4975 SEK
A17268.0E A17268.22 A17268.36
L(+)-Lactic acid ∼85 - 90% aqueous solution
L(+)-Lactic acid
CAS No.: 79-33-4 Molecular Formula: C3H6O3 Formula Weight: 90.08 Flash Point: 110°(230°F) Refractive Index: 1.4270 Odour: Odourless pH: 2.8 Melting Point: 18° Beilstein Reference: 1720251 Merck Reference: 14,5337 Optical Rotation: -13° (c=2.5 in 1N NaOH) MDL No.: MFCD00064266
Formel: C₃H₆O₃
MW: 90.08 g/mol
Kokpunkt: 235
Smältpunkt: 18 °C
Densitet: 1.209
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064266
CAS nummer: 79-33-4
EINECS: 201-196-2
Merck Index: 14,5337
Order Now
SPECIFIKATIONER
Environmentally Preferable