Agaros ME for electrophoresis of macromolecules
Leverantör: Thermo Fisher Scientific
A16693.06 A16693.14
A16693.06EA 785 SEK
A16693.06 A16693.14
Agaros ME for electrophoresis of macromolecules
Agaros
CAS No.: 9012-36-6 MDL No.: MFCD00081294
Smältpunkt: 60…90 °C
Storage Temperature: Omgivande
MDL Number: MFCD00081294
CAS nummer: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM