Cellulose MN 100 adsorbents for column chromatography
Leverantör: MACHEREY-NAGEL
Synonyms: Microcrystallinecellulose
815050.1 815050.25 815050.5
MANA815050.1EA 680 SEK
MANA815050.1 MANA815050.25 MANA815050.5
Cellulose MN 100 adsorbents for column chromatography
Cellulose
Native fibrous cellulose with an average degree of polymerisation 620 to 680.
  • For separation of hydrophilic substances
  • Fiber length (85%): 20 to 100 μm
  • Specific surface according to Blaine ~6500 cm²/g
Formel: (C₁₂H₂₂O₁₁)n
MW: 342.3 g/mol
Smältpunkt: 260…270 °C
Densitet: 1.27…1.6 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00081512
CAS nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977
Order Now
Specifikation testresultat
Residue on Ignition <10000 ppm
Fe <20 ppm
Cu <5 ppm
P <7 ppm
Dichloromethane Extract <0.20%