D (+)-Glukos, vattenfri 99%
Leverantör: Thermo Scientific
Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
A16828.36 A16828.0C A16828.0E
A16828.36EA 665 SEK
A16828.36 A16828.0C A16828.0E
D (+)-Glukos, vattenfri 99%
D (+)-Glukos
Purity: 99% CAS No.: 50-99-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Cryst powder Appearance: White Melting Point: 147-153° Beilstein Reference: 1281608 Merck Reference: 14,4459 Optical Rotation: +53° (c=10 in water, 3h) MDL No.: MFCD00063774
Formel: C₆H₁₂O₆
MW: 180,16 g/mol
Smältpunkt: 147…153 °C
Densitet: 1,54 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00148912
CAS nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,4459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=6f577c634db56f569390df302bc5753ae3fbc654&prid=web&clid=SAPDAM