D (+)-Sackaros ≥99.9%, Ultrapure Bioreagent for density gradient ultracentrifugation, för molekylärbiologi, J.T.Baker®
Leverantör: Avantor
Varning
Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
4097-04EA 0 SEK
4097-04 4097-07
D (+)-Sackaros ≥99.9%, Ultrapure Bioreagent for density gradient ultracentrifugation, för molekylärbiologi, J.T.Baker®
D (+)-Sackaros
Sucrose for Density Gradient Centrifugation and Molecular Biology Applications.
Formel: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Smältpunkt: 169…170 °C
Densitet: 1.29 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006626
CAS nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
For Laboratory,Research,or Manufacturing Use
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
Clarity and Color of Solution Passes Test
Clarity of Solution Passes Test
Glucose ≤0.1 %
Total of Copper, Iron and Lead ≤5 ppm
Assay (C₁₂H₂₂O₁₁) (by HPLC) ≥99.9 %