Bortrifluoriddietyleterkomplex ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A15275.AE A15275.AP A15275.AU
A15275.AEEA 775 SEK
A15275.AE A15275.AP A15275.AU
Bortrifluoriddietyleterkomplex ≥98%
Bortrifluoriddietyleterkomplex
Purity: 98+% CAS No.: 109-63-7 Molecular Formula: C4H10BF3O Formula Weight: 141.93 Flash Point: 47°(116°F) Refractive Index: 1.3440 Odour: Acrid Melting Point: -60° Boiling Point: 125-126° Beilstein Reference: 3909607 Merck Reference: 14,1350 UN No.: 2604 MDL No.: MFCD00013194
Formel: C₄H₁₀BF₃O
Kokpunkt: 125…126 °C
Smältpunkt: –60 °C
Densitet: 1,130
Flampunkt: 47 °C (116 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013194
CAS nummer: 109-63-7
EINECS: 203-689-8
UN: 2604
ADR: 8,I
Merck Index: 14,1350