(C8-18)alkylbenzyldimetylammoniumklorid
Leverantör: Thermo Scientific
Fara
Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
41339.14 41339.22 41339.36
41339.14EA 750 SEK
41339.14 41339.22 41339.36
(C8-18)alkylbenzyldimetylammoniumklorid
(C8-18)alkylbenzyldimetylammoniumklorid
CAS No.: 63449-41-2 Molecular Formula: C6H5CH2N(CH3)2RCl Physical Form: Semi-solid/solid, R=C8H17 to C18H37 Solubility: Soluble in water Merck Reference: 14,1059 UN No.: 2923 MDL No.: MFCD00145757
Cationic surface active agent
Formel: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Smältpunkt: 29…34 °C
Densitet: 0,98 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00145757
CAS nummer: 63449-41-2
EINECS: 264-151-6
UN: 2923
ADR: 8,III
Merck Index: 14,1059