Fullerene C60 ≥99.5%, pulver
Leverantör: Thermo Scientific
Varning
Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen
39722.03 39722.MC
39722.03EA 11200 SEK
39722.03 39722.MC
Fullerene C60 ≥99.5%, pulver
Fullerene C60
Purity: 99.5% CAS No.: 99685-96-8 Density: 1.7 Appearance: Black/brown Boiling Point: 500-600° subl. UN No.: 1325 MDL No.: MFCD00151408
Formel: C₆₀
MW: 720,66 g/mol
Kokpunkt: 500…600 °C (subl.)
Smältpunkt: > 280 °C
Densitet: 1,7
Storage Temperature: Omgivande
MDL Number: MFCD00151408
CAS nummer: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=373518502f0ccac81d515a1161601ce523b24c3f&prid=web&clid=SAPDAM