Molekylsikt 3A (0,3 nm, 3 Å), beads 1 - 2 mm (0.04-0.08 in)
Leverantör: Thermo Scientific
Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
L05335.30 L05335.0B L05335.0I
L05335.30EA 1300 SEK
L05335.30 L05335.0B L05335.0I
Molekylsikt 3A (0,3 nm, 3 Å), beads 1 - 2 mm (0.04-0.08 in)
Molecular sieve 3A (0.3 nm, 3 Å)
CAS No.: 308080-99-1 MDL No.: MFCD00131613
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Omgivande
MDL Number: MFCD00131613
CAS nummer: 308080-99-1