Molekylsikt 3A (0,3 nm, 3 Å), pellets, Ø 3 - 4 mm (0.12 - 0.16 in)
Leverantör: Thermo Scientific
Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
33530.A1 33530.A7
33530.A1EA 2750 SEK
33530.A1 33530.A7
Molekylsikt 3A (0,3 nm, 3 Å), pellets, Ø 3 - 4 mm (0.12 - 0.16 in)
Molecular sieve 3A (0.3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Omgivande
MDL Number: MFCD00131613
CAS nummer: 308080-99-1