1,2-Diaminocyclohexane mixture of isomers 99%
Leverantör: Thermo Scientific
Fara
Synonyms: 1,2-Diaminocyclohexane, 1,2-Cyclohexanediamine
B24657.0B B24657.30
B24657.0BEA 1925 SEK
B24657.0B B24657.30
1,2-Diaminocyclohexane mixture of isomers 99%
1,2-Diaminocyclohexane
Purity: 99% CAS No.: 694-83-7 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 75°(167°F) Refractive Index: 1.4870 Odour: Amine-like pH: 12 Boiling Point: 191° Beilstein Reference: 506142 UN No.: 2735 MDL No.: MFCD00001491
Formel: C₆H₁₄N₂
MW: 114,19 g/mol
Kokpunkt: 191 °C
Smältpunkt: 41…44 °C
Densitet: 0,94
Flampunkt: 75 °C (167 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001491
CAS nummer: 694-83-7
EINECS: 211-776-7
UN: 2735
ADR: 8,II