Isoeugenol cis- and trans-mixture ≥98%
Leverantör: Thermo Fisher Scientific
Varning
B24541.14 B24541.22 B24541.36
B24541.14EA 635 SEK
B24541.14 B24541.22 B24541.36
Isoeugenol cis- and trans-mixture ≥98%
Isoeugenol
Purity: 98+% CAS No.: 97-54-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 134°(273°F) Refractive Index: 1.5770 Melting Point: -10° Boiling Point: 266-268° Beilstein Reference: 1909602 Merck Reference: 14,5171 MDL No.: MFCD00009285
Formel: C₁₀H₁₂O₂
Kokpunkt: 266…268 °C
Smältpunkt: –10 °C
Densitet: 1,080
Flampunkt: 134 °C (273 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009285
CAS nummer: 97-54-1
EINECS: 202-590-7
Merck Index: 14,5171