Kresol ≥99.0%
Leverantör: TCI
|
Fara
|
C0698-500ML
C0698-25ML
TCIAC0698-500MLEA
1625
SEK
TCIAC0698-500ML
TCIAC0698-25ML
Kresol ≥99.0%
Kresol
Formel:
C₇H₈O Kokpunkt: 192°C Densitet: 1,03 g/cm³ (20 °C) Flampunkt: 81°C |
MDL Number:
MFCD00151099 CAS nummer: 1319-77-3 EINECS: 215-293-2 UN: 2022 ADR: 6.1,II Merck Index: 14,02579 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % (Equivalent mixture of o, m, p) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...