Natronkalk, med indikator
Leverantör: Thermo Fisher Scientific
|
Fara
|
44786.36
44786.A4
44786.36EA
3825
SEK
44786.36
44786.A4
Natronkalk, med indikator
Soda lime
CAS No.: 8006-28-8
Physical Form: 8-12 Mesh
Sensitivity: Air Sensitive and Hygroscopic
Odour: Odourless
Merck Reference: 14,8569
UN No.: 1907
MDL No.: MFCD00134094
Storage Temperature:
Omgivande |
MDL Number:
MFCD00134094 CAS nummer: 8006-28-8 UN: 1907 ADR: 8,III Merck Index: 14,8569 |