Platinum ≥99.95%, tråd, hard, Ø 0.6 mm (0.024 in)
Leverantör: Thermo Scientific
Synonyms: Platinium
43494.BS 43494.BW
43494.BSEA 6150 SEK
43494.BS 43494.BW
Platinum ≥99.95%, tråd, hard, Ø 0.6 mm (0.024 in)
Platinum
Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈6.06g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
MW: 195,08 g/mol
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689