Platinum ≥99.95%, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Platinium
40419.KG 40419.KJ
40419.KGEA 16800 SEK
40419.KG 40419.KJ
Platinum ≥99.95%, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Platinum
Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈4.32g/slug Odour: Odourless MDL No.: MFCD00011179
Evaporation source, jewelry production, electrical contacts for semiconductors
Formel: Pt
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689