Platinum ≥99.9%, amorphous powder APS <3 µm
Leverantör: Thermo Fisher Scientific
36704.03 36704.06
36704.03EA 6725 SEK
36704.03 36704.06
Platinum ≥99.9%, amorphous powder APS <3 µm
Platinum
Purity: 99.9% CAS No.: 7440-06-4 Physical Form: S.A. 3.5-7.9m2/g, Tap density 0.6-2.0g/cm3 Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
Kokpunkt: 4530 °C (1013 hPa)
Smältpunkt: 1773 °C
Densitet: 21,4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II