Platinum ≥99.95%, tråd, hard, Ø 1.5 mm (0.059 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Platinium
13153.BP 13153.BS 13153.BW
13153.BPEA 4800 SEK
13153.BP 13153.BS 13153.BW
Platinum ≥99.95%, tråd, hard, Ø 1.5 mm (0.059 in)
Platinum
Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈37.9g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689