Platinum ≥99.95%, tråd, hard, Ø 0.368 mm (0.0145 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Platinium
13039.BU 13039.BY 13039.CC
13039.BUEA 4500 SEK
13039.BU 13039.BY 13039.CC
Platinum ≥99.95%, tråd, hard, Ø 0.368 mm (0.0145 in)
Platinum
Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈2.28g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689