Platinum ≥99.99%, stavar, Premion®, Ø 6.35 mm (0.25 in)
Leverantör: Thermo Scientific
Synonyms: Platinium
12556.DJ 12556.HF
12556.DJEA 0 SEK
12556.DJ 12556.HF
Platinum ≥99.99%, stavar, Premion®, Ø 6.35 mm (0.25 in)
Platinum
Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈6.8g/cm Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
MW: 195,08 g/mol
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689