Platinum ≥99.997%, folie, Premion®, Thickness: 0.1 mm (0.004 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Platinium
12059.FF 12059.FI 12059.GH
12059.FFEA 11175 SEK
12059.FF 12059.FI 12059.GH
Platinum ≥99.997%, folie, Premion®, Thickness: 0.1 mm (0.004 in)
Platinum
Purity: 99.997% CAS No.: 7440-06-4 Physical Form: ≈1.34g/25x25mm Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689