Platinum ≥99.99%, folie, Premion®, Thickness: 0.025 mm (0.001 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Platinium
11510.FF 11510.FI 11510.GH
11510.FFEA 5150 SEK
11510.FF 11510.FI 11510.GH
Platinum ≥99.99%, folie, Premion®, Thickness: 0.025 mm (0.001 in)
Platinum
Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈0.34g/25x25mm Density: 21.45G/CC Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,45G/CC
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689