Platinum ≥99.99%, folie, Premion®, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Scientific
Synonyms: Platinium
11508.FF 11508.FI 11508.GH
11508.FFEA 18300 SEK
11508.FF 11508.FI 11508.GH
Platinum ≥99.99%, folie, Premion®, Thickness: 0.25 mm (0.01 in)
Platinum
Purity: 99.99% CAS No.: 7440-06-4 Physical Form: ≈3.35g/25x25mm Odour: Odourless MDL No.: MFCD00011179
Formel: Pt
MW: 195,08 g/mol
Kokpunkt: 3827 °C (1013 hPa)
Smältpunkt: 1769 °C
Densitet: 21,447 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689