Platinum ≥99.9%, folie, Thickness: 0.635 mm (0.025 in)
Leverantör: Thermo Fisher Scientific
00860.FF
00860.FI
00860.FFEA
35300
SEK
00860.FF
00860.FI
Platinum ≥99.9%, folie, Thickness: 0.635 mm (0.025 in)
Platinum
Purity: 99.9%
CAS No.: 7440-06-4
Physical Form: ≈8.5g/25x25mm
Odour: Odourless
MDL No.: MFCD00011179
Formel:
Pt Kokpunkt: 3827 °C (1013 hPa) Smältpunkt: 1769 °C Densitet: 21,447 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00011179 CAS nummer: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...