Palladium 99,95 Metaller basis, powder APS 0,5-1,7 micron
Leverantör: Thermo Fisher Scientific
Fara
43715.03 43715.14 43715.06
43715.03EA 8000 SEK
43715.03 43715.14 43715.06
Palladium 99,95 Metaller basis, powder APS 0,5-1,7 micron
Palladium
Purity: 99.95% CAS No.: 7440-05-3 Physical Form: Tap density 2.4-5.0g/cm3, S.A. 0.7-2.0m2/g Odour: Odourless UN No.: 3089 MDL No.: MFCD00011167
Formel: Pd
MW: 106.420 g/mol
Densitet: 12.02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1 II