Palladium ≥99.995%, pulver, Premion® , -22 mesh
Leverantör: Thermo Fisher Scientific
Varning
10788.03 10788.06 10788.93
10788.03EA 8200 SEK
10788.03 10788.06 10788.93
Palladium ≥99.995%, pulver, Premion® , -22 mesh
Palladium
Purity: 99.995% CAS No.: 7440-05-3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00011167
Formel: Pd
Densitet: 12,02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,III