Palladium ≥99.95%, thin foil, Thickness: 0,0125 mm (0.0005 in)
Leverantör: Thermo Scientific
40744.FF 40744.FI
40744.FFEA 6400 SEK
40744.FF 40744.FI
Palladium ≥99.95%, thin foil, Thickness: 0,0125 mm (0.0005 in)
Palladium
Purity: 99.95% CAS No.: 7440-05-3 Physical Form: ≈0.094g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00011167
Formel: Pd
MW: 106,420 g/mol
Kokpunkt: 3167 °C (1013 hPa)
Smältpunkt: 1555 °C
Densitet: 12,02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121