Palladium 99,9 Metaller basis, slug, Length: 6.35 mm, Ø 6.35 mm
Leverantör: Thermo Fisher Scientific
40418.KG 40418.KJ
40418.KGEA 19275 SEK
40418.KG 40418.KJ
Palladium 99,9 Metaller basis, slug, Length: 6.35 mm, Ø 6.35 mm
Palladium
Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈2.41g/slug Odour: Odourless MDL No.: MFCD00011167
Evaporation source, hydrogen getter, jewelry production, electrical contacts for semiconductors
Formel: Pd
MW: 106.420 g/mol
Densitet: 12.02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6