Palladium 99,9 Metaller basis, foil, Thickness: 0.5 mm
Leverantör: Thermo Fisher Scientific
11514.FF 11514.FI
11514.FFEA 29275 SEK
11514.FF 11514.FI
Palladium 99,9 Metaller basis, foil, Thickness: 0.5 mm
Palladium
Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈3.8g/25x25mm Odour: Odourless MDL No.: MFCD00011167
Formel: Pd
MW: 106.420 g/mol
Densitet: 12.02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6