Palladium 99,99 Metaller basis, wire, Premion®, Ø 0.5 mm
Leverantör: Thermo Fisher Scientific
10961.BW 10961.CB 10961.CD
10961.BWEA 9425 SEK
10961.BW 10961.CB 10961.CD
Palladium 99,99 Metaller basis, wire, Premion®, Ø 0.5 mm
Palladium
Purity: 99.99+% CAS No.: 7440-05-3 Physical Form: ≈2.36g/m Odour: Odourless MDL No.: MFCD00011167
Formel: Pd
MW: 106.420 g/mol
Densitet: 12.02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6