Palladium 99,9 Metaller basis, wire, Ø 1.0 mm
Leverantör: Thermo Fisher Scientific
10280.BS 10280.BW 10280.CB
10280.BSEA 7550 SEK
10280.BS 10280.BW 10280.CB
Palladium 99,9 Metaller basis, wire, Ø 1.0 mm
Palladium
Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈9.44g/m Odour: Odourless MDL No.: MFCD00011167
Formel: Pd
MW: 106.420 g/mol
Densitet: 12.02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6