Palladium 99,9 Metaller basis, wire, Ø 0.25 mm
Leverantör: Thermo Fisher Scientific
10278.G1 10278.G5
10278.G1EA 4825 SEK
10278.G1 10278.G5
Palladium 99,9 Metaller basis, wire, Ø 0.25 mm
Palladium
Purity: 99.9% CAS No.: 7440-05-3 Physical Form: ≈0.6g/m Odour: Odourless MDL No.: MFCD00011167
Formel: Pd
MW: 106.420 g/mol
Densitet: 12.02 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6