Nickel 99,999 Metaller basis, rods, Puratronic®, Ø 5 mm (0.2 in)
Leverantör: Thermo Fisher Scientific
Fara
42835.DJ 42835.DO
42835.DJEA 7625 SEK
42835.DJ 42835.DO
Nickel 99,999 Metaller basis, rods, Puratronic®, Ø 5 mm (0.2 in)
Nickel
Purity: 99.999% CAS No.: 7440-02-0 Physical Form: ≈1.75g/cm Odour: Odourless MDL No.: MFCD00011137
Formel: Ni
MW: 58.69 g/mol
Kokpunkt: 3007
Smältpunkt: 1453
Densitet: 8.9 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011137
CAS nummer: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582