Nickel ≥99.995%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
Fara
42332.09 42332.18 42332.30
42332.09EA 1650 SEK
42332.09 42332.18 42332.30
Nickel ≥99.995%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Nickel
Purity: 99.995% CAS No.: 7440-02-0 Physical Form: ≈0.45g/slug Odour: Odourless MDL No.: MFCD00011137
Formel: Ni
Kokpunkt: 3007 °C (1013 hPa)
Smältpunkt: 1453 °C
Densitet: 8,9 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011137
CAS nummer: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582