Nickel ≥99.994%, folie, Puratronic®, Thickness: 0.1 mm (0.004 in)
Leverantör: Thermo Scientific
Fara
12046.FI 12046.GH 12046.HP
12046.FIEA 4775 SEK
12046.FI 12046.GH 12046.HP
Nickel ≥99.994%, folie, Puratronic®, Thickness: 0.1 mm (0.004 in)
Nickel
Purity: 99.994% CAS No.: 7440-02-0 Physical Form: ≈2.2g/50x50mm Odour: Odourless MDL No.: MFCD00011137
Formel: Ni
MW: 58,69 g/mol
Kokpunkt: 3007 °C (1013 hPa)
Smältpunkt: 1453 °C
Densitet: 8,9 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011137
CAS nummer: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582