Iridium hard ≥99.8%, folie, hard, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Fisher Scientific
11427.FF 11427.FI
11427.FFEA 61075 SEK
11427.FF 11427.FI
Iridium hard ≥99.8%, folie, hard, Thickness: 0.25 mm (0.01 in)
Iridium
Purity: 99.8% CAS No.: 7439-88-5 Physical Form: ≈3.5g/25x25mm, Temper: as rolled MDL No.: MFCD00011062
Formel: Ir
Kokpunkt: 5300 °C (1013 hPa)
Smältpunkt: 2450 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011062
CAS nummer: 7439-88-5
EINECS: 231-095-9