Kobolt ≥99.995%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Scientific
Fara
43226.QJ 43226.QK
43226.QJEA 3900 SEK
43226.QJ 43226.QK
Kobolt ≥99.995%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Kobolt
Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈1.8g/slug Odour: Odourless MDL No.: MFCD00010935
Formel: Co
MW: 58,93 g/mol
Kokpunkt: 2870 °C (1013 hPa)
Smältpunkt: 1495 °C
Densitet: 8,9 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010935
CAS nummer: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488