Tantalum ≥99.95%, slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Scientific
42826.09 42826.18
42826.09EA 2725 SEK
42826.09 42826.18
Tantalum ≥99.95%, slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Tantalum
Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈0.42g/slug Odour: Odourless MDL No.: MFCD00011252
Formel: Ta
MW: 180,95 g/mol
Kokpunkt: 5427 °C (1013 hPa)
Smältpunkt: 2996 °C
Densitet: 16,69 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011252
CAS nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223