Tantalum 99,9 Metaller basis, rods, Ø 4.75 mm (0.19 in)
Leverantör: Thermo Fisher Scientific
42494.BQ 42494.BU
42494.BQEA 2850 SEK
42494.BQ 42494.BU
Tantalum 99,9 Metaller basis, rods, Ø 4.75 mm (0.19 in)
Tantalum
Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈3.26g/cm Odour: Odourless MDL No.: MFCD00011252
Formel: Ta
MW: 180.95 g/mol
Kokpunkt: 5427
Smältpunkt: 2996
Densitet: 16.69 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011252
CAS nummer: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223