Niob ≥99.99%, pulver, Puratronic® -325 mesh (max. 500ppm Ta)
Leverantör: Thermo Fisher Scientific
Fara
40510.09 40510.18
40510.09EA 2125 SEK
40510.09 40510.18
Niob ≥99.99%, pulver, Puratronic® -325 mesh (max. 500ppm Ta)
Niobium
Purity: 99.99% CAS No.: 7440-03-1 Physical Form: APS 10 micron Odour: Odourless UN No.: 3089 MDL No.: MFCD00011126
Formel: Nb
MW: 92.91 g/mol
Kokpunkt: 3700
Smältpunkt: 2468
Storage Temperature: Omgivande
MDL Number: MFCD00011126
CAS nummer: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II