Niob ≥99.95%, slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
42846.09 42846.18
42846.09EA 1275 SEK
42846.09 42846.18
Niob ≥99.95%, slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Niobium
Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈0.43g/slug Odour: Odourless MDL No.: MFCD00011126
Formel: Nb
Kokpunkt: 3700 °C (1013 hPa)
Smältpunkt: 2468 °C
Densitet: 8,57 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011126
CAS nummer: 7440-03-1
EINECS: 231-113-5