Zirconium ≥99.2%, plåt 100x100 mm (3.9x3.9 in), Thickness: 6.4 mm (0.25 in)
Leverantör: Thermo Scientific
44261.KF 44261.KJ
44261.KFEA 6325 SEK
44261.KF 44261.KJ
Zirconium ≥99.2%, plåt 100x100 mm (3.9x3.9 in), Thickness: 6.4 mm (0.25 in)
Zirconium
Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈416g/100x100mm, Temper: annealed Odour: Odourless MDL No.: MFCD00011303
Formel: Zr
MW: 91,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011303
CAS nummer: 7440-67-7
EINECS: 231-176-9