Zirconium ≥99.8%, folie (Hf ∼ 2%), annealed, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Scientific
10591.GI 10591.GR 10591.IB
10591.GIEA 1400 SEK
10591.GI 10591.GR 10591.IB
Zirconium ≥99.8%, folie (Hf ∼ 2%), annealed, Thickness: 0.25 mm (0.01 in)
Zirconium
Purity: 99.8% CAS No.: 7440-67-7 Physical Form: ≈20.6g/100x125mm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Formel: Zr
MW: 91,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011303
CAS nummer: 7440-67-7
EINECS: 231-176-9